2-Acetylamino-5-bromo-4-methylpyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA2823-1GEA 435 DKK
TCIAA2823-1G
2-Acetylamino-5-bromo-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Acetylamino-5-bromo-4-methylpyridine
Formel: C₈H₉BrN₂O
MDL Number: MFCD06798092
CAS-nummer: 142404-82-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR