(R)-2-Amino-1-phenylethanol ≥96.0% (by HPLC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA2489-1GEA
517
DKK
TCIAA2489-1G
(R)-2-Amino-1-phenylethanol ≥96.0% (by HPLC)
(R)-2-Amino-1-phenylethanol
Formel:
C₈H₁₁NO Molekylvægt: 137,18 g/mol |
MDL Number:
MFCD00239406 CAS-nummer: 2549-14-6 |
Specifikationer Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 58.0 to 64.0 deg-C |
Specific rotation [a]20/D | -39.0 to -45.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...