2-Chloroethyl acetate ≥99.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAA0027-25MLEA
788
DKK
TCIAA0027-25ML
2-Chloroethyl acetate ≥99.0% (by GC)
2-Chloroethyl acetate
Formel:
C₄H₇ClO₂ Kogepunkt: 145°C Flammepunkt: 54°C |
MDL Number:
MFCD00018963 CAS-nummer: 542-58-5 EINECS: 208-820-2 UN nummer: 2929 ADR: 6.1,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...