Disposable particle filtering respirator, FFP3, Aura™ 9332+
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Aura™
111-2533EA 20450 DKK
111-2533
Disposable particle filtering respirator, FFP3, Aura™ 9332+
Åndedrætsværn Engangsmasker Partikelmasker
3M™ Aura™ Particulate Respirator 9332+ provides FFP3 breathing protection against dusts, mists, and metal fumes in a wide variety of industrial applications and other hazardous environments.
  • Original 3-panel design allows for greater facial movement
  • Low-resistance filter technology facilitates easy breathing
  • Cool Flow™ valve reduces heat build-up in hot and humid conditions
  • Sculpted upper panel conforms to the nose and contours of the face for an effective seal
  • Embossed top panel reduces eyewear fogging
  • Easy to store away with flat-fold design and individual wrapping
  • Colour-coded headbands for easy identification: red for FFP3 (NFP 50)
  • Maximum usage level: Up to 50 x TLV
The 3-panel design allows for greater facial movement, while the sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging, and improves eyewear compatibility.
This model features the 3M™ Cool Flow™ valve, which reduces heat build-up in hot and humid conditions, and the low-resistance filter technology permits easy breathing.
Certificeringer: EN 149:2001+A1:2009 FFP3 NR D
Emballering: This respirator is individually packed in hygienic wrapping to protect against contamination.