Disposable particle filtering respirator, FFP2, Aura™ 9322+
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Aura™
MMMA9322PLUSBVEA 9390 DKK
MMMA9322PLUSBV
Disposable particle filtering respirator, FFP2, Aura™ 9322+
Åndedrætsværn Engangsmasker Partikelmasker
3M™ Aura™ Particulate Respirator 9322+ provides FFP2 breathing protection with a flat-fold, 3-panel design, low breathing resistance and an embossed, sculpted top panel that reduces eyewear fogging. It features an innovative chin tab for easy adjustments, and the headbands are colour-coded for quick and easy identification.
  • Original 3-panel design allows for greater facial movement
  • Low resistance filter technology facilitates easy breathing
  • Sculpted upper panel conforms to the nose and contours of the face for an effective seal
  • Cool Flow™ valve reduces heat build-up in hot and humid conditions
  • Embossed top panel reduces eyewear fogging
  • Easy to store away with flat-fold design and individual wrapping
  • Colour-coded headbands for easy identification: blue for FFP2 (NFP 12)
Get FFP2 protection against dusts and mists in a wide variety of industrial applications and other hazardous environments with 3M™ Aura™ Particulate Respirator 9322+. The 3-panel design allows for greater facial movement, while the sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging, and improves eyewear compatibility.
This model features the 3M™ Cool Flow™ valve, which reduces heat build-up in hot and humid conditions, and the low-resistance filter technology permits easy breathing. The flat-fold design makes it convenient to store when not in use, and the innovative chin tab allows simple adjustments. The braided, colour-coded headbands help to identify the protection level offered: blue for FFP2 (NPF 12).
This respirator is individually packed in hygienic wrapping to protect against contamination, and it’s designed to work well with 3M Eyewear and Hearing Protection products.