tetra-Chlorauric (III)syre 2% i vandigopløsning
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
190493PEA
0
DKK
190493P
tetra-Chlorauric (III)syre 2% i vandigopløsning
tetra-Chlorauric (III)syre
2% w/v HAuCl₄.3H₂O
Formel:
HAuCl₄ Molekylvægt: 339,78 g/mol |
MDL Number:
MFCD00011322 CAS-nummer: 16903-35-8 EINECS: 240-948-4 UN nummer: 3264 ADR: 8,III |
Specifikationer Testresultater
Assay (gravimetrically) (W/V) | 1.98 - 2.02 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...