Du Søgte efter: Fuldtdækkende ansigtsmasker
16  results were found
SearchResultCount:"16"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: 3M
Beskrivelse: The 3M™ Secure Click™ full facepiece reusable respirator FF-800 series helps provide protection against certain airborne contaminants. For use with 3M...
New Product
Leverandør: JSP LTD
Beskrivelse: The Force®10 Typhoon™ is an extremely lightweight, easy to maintain, full face mask with panoramic visor. With 3 available sizes (S,M,L) and the abili...
Tilbud
Varenummer: (MMMASK102)
Leverandør: 3M
Beskrivelse: Wear prescription lenses while using certain 3M™ Reusable Full Face Masks with the help of 3M™ Reusable Respirator Spectacle Kit. These spectacle...
UOM: 1 * 1 Each
Leverandør: 3M
Beskrivelse: Letvægts hel ansigtsmasker for maksimal komfort. Inkorporerer et blødt, hypoallergent materiale og central udåndingsventil.
Leverandør: 3M
Beskrivelse: 3M™ Full Facepiece Reusable Respirator is multi-function full-face mask designed to provide durable and comfortable protection against a wide range of...
New Product
Leverandør: 3M
Beskrivelse: 3M™ reusable full face mask, FF-600 series features a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M...
New Product
Varenummer: (PRTWP516BLU)
Leverandør: Portwest
Beskrivelse: The Helsinki full face mask is made with a soft thermoplastic rubber face seal and features a panoramic viewer.
UOM: 1 * 1 Items
Leverandør: Dräger
Beskrivelse: X-plore® 5500 helmasken er den ideelle løsning til alle arbejdsopgaver, som kræver åndedrætsværn, f.eks. i den kemiske industri, i metal- og bilindust...
Leverandør: Dräger
Beskrivelse: X-plore® 6530-modellen er udstyret med åndedrætsfiltre fra den omfattende Dräger X-plore® Rd40-filterserie og skiller sig dermed ud ved at være egnet ...
Leverandør: Moldex
Beskrivelse: Extremely lightweight, reusable full face mask (only 360 g) made from soft, TPE material.
Sikrere sundhed for mennesker og miljø
Varenummer: (111-1901)
Leverandør: Dräger
Beskrivelse: X-plore® 6570 er den silikonehelmaske med høj komfort, der anvendes af fagfolk til en bred vifte af anvendelser. Til anvendelser med ukendte smitstoff...
UOM: 1 * 1 Items
Varenummer: (111-0777)
Leverandør: Dräger
Beskrivelse: The reusable X-plore® 6300 is the successor to the Panorama Nova Standard – redesigned and improved with fresh colours and an integrated bar code.
UOM: 1 * 1 Items
Leverandør: HONEYWELL SAFETY
Beskrivelse: Optifit is a reusable, APR class 2 full face mask dedicated to offer respiratory protection, depending on the filter type assembled on it. For optimal...
Leverandør: HONEYWELL SAFETY
Beskrivelse: Twin filter masks give general protection against particulates and/or toxic gases. They can only be used in sufficiently ventilated premises where the...
Varenummer: (111-3060)
Leverandør: BARTELS + RIEGER GMBH
Beskrivelse: This reusable full face mask is for use in industry, agriculture, rescue work and the disposal of hazardous substances. In addition to its use as a st...
UOM: 1 * 1 Items
Varenummer: (111-1455)
Leverandør: HONEYWELL SAFETY
Beskrivelse: Panoramasque is a full face mask dedicated to offer respiratory protection, depending on the canister type assembled on it. For optimal protection, it...
UOM: 1 * 1 Items
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 16
no targeter for Bottom