Density meter, Ultrospec™ 10 Classic
Leverandør: Biochrom
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Ultrospec™
634-0882EA 10730 DKK
634-0882
Density meter, Ultrospec™ 10 Classic
Massefylde målere
The Ultrospec 10 Classic can measure growth rates of Escherichia coli, other bacteria and yeast in suspension at 600 nm.
  • Spillages wiped from the smooth surface
  • Cell compartment area can be sterilised
  • Download results to a PC for convenient data storage and retrieval
For small research labs, the density meter must be taken where the cells are grown and used either in incubation cabinets or under anaerobic conditions.
Advarsel: Not recommended for use with mammalian cell lines.