Very powerful workshop cream, to remove traces of paint, ink, resin and glue, as well as most of the other contaminating agents present in printing and painting workshops.
- Natural charges of non-abrasive corn cleans the skin thoroughly without damaging it
- Fruity fragrance leaves afeeling of freshness on the skin
- Based on ester dibasic, agent recognised for its highly cleaning action
- Non-irritant - 48-hour dermatological patch test
Its cleaning power comes from the combination of dibasic esters and natural corn-based fillers. Contains a moisturising agent so that its regular use does not affect the hydration of the skin. Waterless formula composed of 100% active material, guaranteeingoptimal cleaning efficiency. Specially designed to achieve a high level of professional efficiency.
Contains a large amount of glycerin,humectant and moisturiser to prevent skin dryness. The Insensitive Loss testWater (PIE) demonstrates that regular use and daily does not dehydrate the skin. Contains extracts of Jojoba and wheat germ which is a source of vitamin E and they allow to leave the soft and supple skin after application.
Applications include heavy industry, general and light industry.