Immersion oil Type N acc. to ISO 8036 for microscopy, Sigma-Aldrich®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.03699.0100EA 825 DKK
1.03699.0100
Immersion oil Type N acc. to ISO 8036 for microscopy, Sigma-Aldrich®
Farvestoffer Mikroskopi og farvningsreagenser
This high quality, halogen-free standard immersion oil provides a defined refractive index and excellent homogeneity to considerably increase lens effectiveness by eliminating refractive distortion, according to the International Standard ISO 8036.
  • Capcity 2500 slides / 100 ml
ISO certification assures consistent performance in terms of optical properties, reference temperature, auto fluorescence, spectral range, density, viscosity and composition. Our immersion oil has been successfully used for decades with some of the world’s highest quality oil immersion objective lenses, without any negative impact on optical quality.
Advarsel: Store at 15 to 25 °C.