Strontium standard for IC, TraceCERT®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SUPL42151-100MLEA 278 DKK
SUPL42151-100ML
Strontium standard for IC, TraceCERT®, Supelco®
Certificeringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)