Cerium Standard for AAS, TraceCERT®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TraceCERT®
53378-250MLEA 195 DKK
53378-250ML
Cerium Standard for AAS, TraceCERT®, Supelco®
Standarder Cationic Standards Cerium Standards
Certificeringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)