Nitrogen generator, Genius NM3G
Leverandør: Peak Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PEAK10-8030EA 0 DKK
PEAK10-8030
Nitrogen generator, Genius NM3G
Gasgeneratorer Nitrogengeneratorer
Specifically engineered for time-critical and emergency applications where an on-site nitrogen generation system which is guaranteed to perform at all times. NM3G nitrogen generation system was designed to provide a variety of extra precautionary features, including highly robust integrated compressors with additional load capacity.
  • Uninterrupted supply of gas for clinical applications
  • Additional compressor capacity ensures continuous operation
  • Advanced alarms notifies product issues
  • Insulated compartment to reduce vibration
  • 1 year comprehensive on-site warranty
Outlet pressure: 6,89 bar
StartUp Time: 30 mins
Power consumption: 2185 W
Weight: 193 kg
Noise level: 54 dB
Certificeringer: Conforms with: Low Voltage Directive 2006/95/EC
EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2004/108/EC
EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
FCC47 CFR Part 15 Class B
Unintentional radiators; Conducted and Radiated emissions limits