Mini power supply, EV0220
Leverandør: Consort
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CONSEV0220EA 0 DKK
CONSEV0220-CH CONSEV0220-UK CONSEV0220
Mini power supply, EV0220
Strømforsyninger
The EV0220 is entry level small power supply suitable for most small tanks and applications. The front panel and graphical display are designed for ease of use. The display provides all useful information during runs and will show an on screen help to guide the user in setting up the power supply.
  • On screen help in four languages to assist the user in setting up the power supply
  • Simple running mode: set voltage, current, power and time
  • Automatic cross-over has constant voltage, constant current, constant power
  • Automatic recovery after
  • Automatic recovery after power failure
  • Password protection
  • Ground leakage detection
The simple mode you just have to set your power supply to the desired parameters and press run. This makes the power supply ideal for teaching purposes. Smooth voltage rise and no load detection.
The complete EV series can keep the voltage constant at low currents without problem and will keep on functioning at low and high temperatures. Consort power supplies are the most robust, long lasting and durable electrophoresis power supplies in the market.