VWR® SMART5, Gel Documentation System
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
730-0496EA 45000 DKK
730-0496
VWR® SMART5, Gel Documentation System
Fotodokumentationssystemer
SMART5 is a new generation of low cost, integrated imaging system for DNA and protein analysis, also gel documentation.
  • Compact workstation
  • Complete with built-in processor, touch screen and SMART5 software
  • Motor-driven zoom lens
  • 5 MP camera
  • White, UV and blue lighting options
  • Complete networking connectivity (optional)
SMART5 is the ultimate in compact gel documentation workstations. Using a choice of UV, blue or white lighting options, you can capture and edit images of fluorescent ethidium bromide, SYBR® Safe or GelGreen™ DNA gels and protein gels stained with Coomassie Blue at the workstation.
For an entry level system, the SMART5 comes with a ground breaking 5 MP CCD camera giving superb pixel resolution and unrivalled sensitivity. SMART5 offers a maximum viewing area of 20×24 cm which is very large for such a small, compact unit.
Leveringsinformation: Complete system includes 5 MP camera with motor-driven zoom lens f1.2; epi white light for sample positioning; UV032 filter; image capture and GeneTools analysis software. Transilluminator and converter screens not supplied (available separately).