Welding spectacles, uvex futura
Leverandør: Uvex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
uvex futura
UVEX9180.145EA 184 DKK
UVEX9180.145 UVEX9180.143
Welding spectacles, uvex futura
Briller
These classic welding protection feature new filter technology for UV and IR radiation protection.
  • Perfect colour recognition thanks to uvex sunglare filters
  • Minimises damage from welding sparks
  • Multi-level side arm inclination and four different side arm lengths adjust to suit any facial shape
  • Soft duo-flex ear pieces for a pressure free fit
Lens coating of uvex infradur with grey welding protection tinting.
Certificeringer: EN 166, EN 169