Powder pots, with lid, stainless steel
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SAMPA166-750EA 5260 DKK
SAMPA166-750 SAMPA166-250 SAMPA166-500 SAMPA166-1000
Powder pots, with lid, stainless steel
These powder pots are wide mouth containers that are ideal for handling powders and granules. They are made from high quality 316L stainless steel and have a hygienic crevice free interior.
  • Robust construction and GMP design
  • High quality mirror polish
  • Satin finish
  • Silicone gasket (FDA accepable)
  • BSE/TSE free
  • Ideal for pharmaceutical, food and chemical companies
The threaded lid is completely removable which means cleaning inside is simple and fast.