Microsart AMP coating buffer
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SARTSMB95-2002EA 723 DKK
SARTSMB95-2002
Microsart AMP coating buffer
The Microsart AMP Coating Buffer is an essential requirement to perform a test for mycoplasma according to the European Pharmacopoeia 2.6.7 with volumes ranging from 0,2 to 18 ml. Mycoplasma particles, which might be present in the sample, are concentrated with Vivaspin® centrifugation units after DNA extraction as part of the sample preparation procedure. The Microsart AMP Coating Buffer is required to condition the sample matrix for use with the filtration membrane.