Twist lock self locking carabiner, DBI-SALA Protecta®, 3M™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DBI-SALA®
CPSDAJ514EA 181 DKK
CPSDAJ514
Twist lock self locking carabiner, DBI-SALA Protecta®, 3M™
Faldbeskyttelse Karabinhager
DBI-SALA® Protecta® Twist lock self locking carabiner features with automatic gate closing and locking which makes this as an ideal choice when the user will open and closed the connector many times during the work period.
  • Quick and easy to open with one hand
  • Made from stainless steel
Certificeringer: EN 362