Buffered sodium chloride peptone solution, with 1% Tween® 80
Leverandør: Millipore
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HEIP1466510006EA 0 DKK
HEIP1466510006
Buffered sodium chloride peptone solution, with 1% Tween® 80
Buffered sodium chloride peptone solution, with 1% Tween® 80, can be used in the microbial examination of non sterile pharmaceutical products.
Emballering: Packed 1 l in 1 l bottle with blue screw cap and 3 loci.