Magnetic isolation device, Mag-ID
Leverandør: OZ Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OZBIDM30000EA 0 DKK
OZBIDM30000
Magnetic isolation device, Mag-ID
Stativer Magnetisk Separation Racks
Magnetic isolation device is ideal for coupling reactions, washing procedures, aspiration, pipetting etc. MagID is made from an injection moulded plastic housing incorporating a high-energy neodymium magnet. It is designed to accommodate standard 1,5 ml tubes and is also suitable for some 2 ml tubes.
  • Ideal for magnetic nanoparticles coupling reaction and purification
  • Adapted to working solutions ranging from 10 µl to 2 ml
  • Perfect for a quick magnetic separation process (<5 minutes) with a high yield separation
  • Durable and easy to use with an open faced design