VWR®, Measuring Cylinders, Class A, TPX
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
612-5841EA 329 DKK
612-5841 612-5842 612-5840 612-5838 612-5839 612-5845 612-5843 612-5844
VWR®, Measuring Cylinders, Class A, TPX
Cylindere Målecylindre
Made from TPX , 100 % virgin resin, clear.
  • Raised graduation and ring marks at the primary scale points
  • Good direct reading by linear meniscus
  • Calibrated ‘In’ at a reference temperature of 20 °C
  • Up to 121 °C (autoclaving) does not cause tolerance limits to be permanently exceeded
  • Hexagonal base with bottom studs to ensure leveling and high stability
  • Product consists engraved batch number
The lot certificate supplied bears the batch number and the actual nominal value ascertained under the test conditions.
The resulting deviations from the nominal value fall well under the allowed tolerances of Class A according to ISO 6706.
Certificeringer: Compliance FDA 21 CFR