VWR®, Serological Pipettes, Open End
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
612-5861EA 1070 DKK
612-5861 612-5862 612-5860 612-5858 612-5859 612-5856 612-5857 612-5855
VWR®, Serological Pipettes, Open End
Pipetter Serologiske pipetter
PS, transparent, graduated, non sterile or sterile. These disposable pipettes for liquid transfers are made of GPPS (general polystyrene), in compliance with USP Class VI.
  • DNase-/RNase-free, non pyrogenic, non cytotoxic, non haemolytic
  • Meet the requirement of BST/TSE; latex-free
  • Graduations are calibrated for accurate dispensing to within ±2%
  • Pipettes are colour coded according to their volume
  • Sterilised by irradiation
  • Every product is printed with a batch number for traceability
Manufactured in a Class 100 000 cleanroom environment. Manufactured under EN ISO 13485 and ISO 9001 quality management systems.