Hexaethyldisiloxan ≥99%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
|
L04367.06EA
650
DKK
L04367.06
Hexaethyldisiloxan ≥99%
Hexaethyldisiloxane
Formel:
C₁₂H₃₀OSi₂ Molekylvægt: 246,54 g/mol Kogepunkt: 231 °C Smeltepunkt: –115 °C Massefylde: 0,840 Flammepunkt: 87 °C (188 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00026693 CAS-nummer: 994-49-0 EINECS: 213-619-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...