Conical sleeves for filtration
Leverandør: Saint-Gobain
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
511-0873EA 368 DKK
511-0873
Conical sleeves for filtration
Beslag Støtteærmer
These rubber filter cones are designed to fit any Büchner funnel, vacuum filter vial or flask, regardless of size. These filter cones can be used with filters, microfilters, porcelain filters and crucibles.
  • Depression ensures contact between the cone and its support
  • The cone acts as a joint not as a stopper