Calciumcarbonat, SAFC®
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.02071.9050EA
0
DKK
1.02071.9050
Calciumcarbonat, SAFC®
Calcium carbonate
Formel:
CaCO₃ Molekylvægt: 100,09 g/mol Smeltepunkt: 825 °C Massefylde: 2,71 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010906 CAS-nummer: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...