1,4-Dihydroxy-9,10-anthraquinon 96%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
ACRO132181000EA
196
DKK
ACRO132181000
ACRO132180010
ACRO132180050
1,4-Dihydroxy-9,10-anthraquinon 96%
1,4-Dihydroxy-9,10-anthraquinon
Formel:
C₁₄H₈O₄ Molekylvægt: 240,22 g/mol Kogepunkt: 465 °C (1013 hPa) Smeltepunkt: 191…193 °C Flammepunkt: 222 °C |
MDL Number:
MFCD00001209 CAS-nummer: 81-64-1 EINECS: 201-368-7 UN nummer: 3077 Merck Index: 12,08250 |
Specifikationer Testresultater
Appearance | Orange to red-brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
Elemental Analysis | 7.5 to 7.9 % N |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...