3-(Diethylamino)phenol 99%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
ACRO146631000EA
313
DKK
ACRO146631000
3-(Diethylamino)phenol 99%
3-(Diethylamino)phenol
Formel:
C₁₀H₁₅NO Molekylvægt: 165,24 g/mol Kogepunkt: 170 °C (15 mmHg) Smeltepunkt: 70…73 °C Flammepunkt: 141 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00002265 CAS-nummer: 91-68-9 EINECS: 202-090-9 |
Specifikationer Testresultater
Appearance | Pink to red to grey-brown Flakes or granules |
Infrared spectrum | Conforms |
Melting point | 70 to 73 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...