Dichlobenil, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,6-Dichlorbenzonitril
M-Clarity™ quality level = MQ100
45431-250MGEA
352
DKK
45431-250MG
Dichlobenil, Supelco®
Dichlobenil
Formel:
C₇H₃Cl₂N Molekylvægt: 172,01 g/mol Kogepunkt: 279 °C (1013 hPa) Smeltepunkt: 145…146 °C Massefylde: 1,623 g/cm³ (–101 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001781 CAS-nummer: 1194-65-6 EINECS: 214-787-5 UN nummer: 3276 ADR: 6.1,III Merck Index: 13,03068 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...