VWR®, Peaked Caps
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
113-8907EA 530 DKK
113-8907 113-8906
VWR®, Peaked Caps
Engangsbeklædning
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 40 g/m² (top), 100 g/m² (peak)
Emballering: 100 pieces per bag, 10 bags per case.