Cobalt(II)acetat tetrahydrat, EMSURE® for analysis, Supelco®
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M-Clarity™ quality level = MQ300
1.02529.0100EA
2380
DKK
1.02529.0100
Cobalt(II)acetat tetrahydrat, EMSURE® for analysis, Supelco®
Cobalt(II)acetat tetrahydrat
Formel:
(H₃CCOO)₂Co·4H₂O Molekylvægt: 249,08 g/mol Smeltepunkt: 140 °C Massefylde: 1,7 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149569 CAS-nummer: 6147-53-1 EINECS: 200-755-8 UN nummer: 3077 ADR: 9,III Merck Index: 12,02494 |
Specifikationer Testresultater
Solubility | 380 g/l (25 °C) |
Melting Point | 140 °C Elimination of water of crystallisation |
Molar Mass | 249.08 g/mol |
Bulk Density | 850 kg/m3 |
Density | 1.70 g/cm3 (20 °C) |
pH | 7.2 (50 g/l, H2O, 20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...