Cleanroom mirrors, CRM
Leverandør: Tinman
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TINMCRM59014802SSEA 4890 DKK
TINMCRM59014802SS TINMCRM5907402SS
Cleanroom mirrors, CRM
These wall mounted cleanroom mirrors complement the gowning area of the cleanroom.
  • Wall mounted, easy assembly
  • Smooth surface: no retention of dirt, contamination free
  • Easy handling, compact size, reduced weight
  • Minimization of gaps, hygienic design
  • Good clean ability due to low surface roughness and easy cleaning access
  • Very good resistance against chemicals and all regular cleanroom disinfectants (alcoholic, biocidal and sporicidal)
Fabrication material: AISI 304 - 8R high gloss polished
Certificeringer: Cleanroom suitability: ISO class 4, GMP Class A