Adjustable sample dividers
Leverandør: ZARGES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ZARG40731EA 1970 DKK
ZARG40731
Adjustable sample dividers
Adjustable sample divider made up of a foam ring and base panel in PE foam ethafoam 220.
  • Nubbly foam for lid, self-adhesive
  • Compartment set made from Kömatex
  • Sheet thickness 5 mm
  • Size of the compartments is approximately 170×170 mm
Unit consists of two lengthwise and three crosswise dividers.
Dimensions: 750×550×330 mm