Crosslinking agents, Sulfo-SMCC
Leverandør: BioVision
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIOV2320-100EA 2280 DKK
BIOV2320-100 BIOV2320-1000 BIOV2320-1EA BIOV2320-50
Crosslinking agents, Sulfo-SMCC
Proteinkoblings- og mærkningssystemer
Sulfo-SMCC is a water-soluble heterobifunctional protein crosslinker with amine reactive Sulfo-NHS ester that crosslinks efficiently with primary amine-containing molecules. It has a maleimide functional group that can be utilised to react specifically with cysteines or sulfhydryl (-SH) groups. Unlike other maleimide-activated antibodies, those made with Sulfo-SMCC crosslinkers are often more stable, due to the inductive stabilising effects of the cyclohexyl ring moiety.
  • White to off-white solid
  • To be protected from moisture
  • Can be used under physiologic conditions
  • Has a spacer arm length of 8,3 Å
Used to prepare hapten-carrier protein and antibody enzyme conjugates, such as HRP conjugation. Also used for both small molecule reactions and large biomolecule applications.
Advarsel: For research use only.