Tungsten ≥99.999% (metals basis), pulver, Puratronic® APS 4-6 µm (max. 1ppm Mo)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

39691.04EA 799 DKK
39691.04
Tungsten ≥99.999% (metals basis), pulver, Puratronic® APS 4-6 µm (max. 1ppm Mo)
Tungsten
Formel: W
Molekylvægt: 183,84 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011461
CAS-nummer: 7440-33-7
EINECS: 231-143-9
UN nummer: 3089
ADR: 4.1,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR