Tantal, sputtering target, Thickness: 3,18 mm
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40894.KSEA
8640
DKK
40894.KS
Tantal, sputtering target, Thickness: 3,18 mm
Tantalum
Formel:
Ta Molekylvægt: 180,95 g/mol Kogepunkt: 5427 °C (1013 hPa) Smeltepunkt: 2996 °C Massefylde: 16,69 g/cm³ (20 °C) |
MDL Number:
MFCD00011252 CAS-nummer: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...