Glassy carbon, spherical powder 1000-2000 µm Type 1

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

41498.18EA 1440 DKK
41498.18
Glassy carbon, spherical powder 1000-2000 µm Type 1
Glassy carbon
Formel: C
Molekylvægt: 12,01 g/mol
Smeltepunkt: 3652 °C
Massefylde: 1,95 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133992
CAS-nummer: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR