Aluminium ≥99% (metals basis), folie [Si & Fe (combined) ∼ 1%], annealed, Thickness: 1.5 mm (0.06 in)
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
44490.HPEA
3770
DKK
44490.HP
Aluminium ≥99% (metals basis), folie [Si & Fe (combined) ∼ 1%], annealed, Thickness: 1.5 mm (0.06 in)
Aluminium
Formel:
Al Molekylvægt: 26,98 g/mol Kogepunkt: 2327 °C (1013 hPa) Smeltepunkt: 660 °C Massefylde: 2,7 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00134029 CAS-nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...