Intelligent battery chargers, Jupiter™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Jupiter™
111-2154EA 3150 DKK
111-2154 111-2369
Intelligent battery chargers, Jupiter™
Batterier
Intelligent charger that cuts off the charging process when battery is fully charged. For use with the Jupiter™ powered air respirator battery pack.