Powder weighing cabinets, Excel Plus Safechange, Bigneat
Leverandør: Bigneat
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Excel Plus
BIGN1606EXCELPLHEPEA 115930 DKK
BIGN1606EXCELPLHEP BIGN1006EXCELPLHEP BIGN1306EXCELPLHEP
Powder weighing cabinets, Excel Plus Safechange, Bigneat
Stinkskabe og kabinetter Ekstraktionsskabe
Excel Plus workstations are used in the laboratory for the most demanding powder handling applications, such as high accuracy weighing, processing of pellets, capsules or tablets and for dispensing of potentially toxic powdered compounds. Construction: All steel parts are epoxy coated; transparent panels are made from clear acrylic which is resistant to corrosion.
  • True Safechange Technology eliminates the possibility of contamination either in the laboratory or upstream in ductwork and fan assembly
  • Double layer filtration provides extra filter system integrity
  • Visible and audible alarms alert user of unsafe flow conditions
  • Fan speed compensation for variation in flow during the lifetime of the filter maintains cabinet performance
  • Double hinged front visors can be opened for full access to the cabinet interior
  • Gentle non-turbulent flow prevents balance fluctuation and cabinet dead spots
  • 180 mm height front access across full cabinet width
Excellent operator protection is achieved by enhanced airflow control and aerofoil technology which ensures that containment is achieved and maintained at the very low air flows required for balance stability.
Two-stage filtration is used in this workstation. A dual set of filters comprising a pre-filter (eliminates particles 5,0 µm or larger to an efficiency of 92% as defined in BS EN ISO 779) then a high capacity design (double layer) HEPA filter (H14 Standard) eliminates particles 0,3 microns or larger to an efficiency of 99,995%.
Power supply: 230 V, AC, 50 Hz, 5 Amp, 1 Ø (on request 110 V, AC, 60 Hz, 8 Amp, 1 Ø).
Leveringsinformation: Supplied with base tray, sealed fluorescent light units, pre-filters, 2× double-layer HEPA filters (Grade H14).