Cleanroom benches, Micro CRB B and Pharma CRB B models, TINMAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TINMMBBT1C1R2D260XEA 5090 DKK
TINMMBBT1C1R2D260X TINMPBBT1C3R2D220X TINMPBBT1C3R2D110X TINMPBBT1C2R2D1800 TINMPBBT1C1R2D1600 TINMPBBT1C4R2D112X TINMMBBT1C3R2D220X TINMMBBT1C2R2D1800 TINMMBBT1C3R2D110X TINMMBBT1C4R2D112X TINMMBBT1C2R2D115X TINMMBBT1C3R2D218X TINMPBBT1C2R2D115X TINMPBBT1C3R2D218X
Cleanroom benches, Micro CRB B and Pharma CRB B models, TINMAN
Theses CRB B benches with bootie racks are designed to assist in putting shoe covers and are ideal in all gowning areas.
  • Ergonomic design: rounded front and back edge – higher comfort level
  • Smooth edges prevents tearing and cuts
  • With perforations: no retention of cleaning fluids ; air permeability 40 % (only Micro Micro CRB B models)
  • Minization of gaps: closed circuit welded joints ; preventing accumulation of dust
  • Good clean ability due to low surface roughness and easy cleaning access
  • Very good resistance against chemicals and all regular cleanroom disinfectants (alcoholic, biocidal and sporicidal)
  • Simple assembly
Two different models are available, the Micro CRB B model with a perforated bench top and the Pharma CRB B model a smooth bench top with its good clean ability due to low surface roughness and easy cleaning access.
Material: AISI 304, 2R - high gloss polished (optional AISI 316)
Cleanrrom sustaibility: ISO class 4, GMP class A