Earmuffs, Peltor™ WS™ Alert™ XP
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MMMAMRX21AWS5EA 5810 DKK
MMMAMRX21AWS5
Earmuffs, Peltor™ WS™ Alert™ XP
These earmuffs feature a built-in FM-radio receiver, Bluetooth® and a level dependent function . The Peltor WS Alert XP enable enables users to have wireless telephone conversations or listen to their favourite music even in noisy environments.
  • FM radio receiver (2G circuit)
  • Wireless calls and music with Stereo Bluetooth®
  • Patented speaker system for full-bodied dynamic sound
  • Super fast tuning and memory/preset function
  • Level dependent function for ambient listening
  • Sound limiter and dosimeter
  • Ghost voice guided interface and configure menu
Stainless steel headband.
Leveringsinformation: Headset, 2 AA batteries (non re-chargeable), user instructions, 1×3,5 mm cable, quick start guide.