Du Søgte efter: Anaerobe systemer
16  results were found
SearchResultCount:"16"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: Merck
Beskrivelse: Anaerocult®-systemet er et pålideligt og anerkendt system til dyrkning af obligat og fakultativt anaerobe organismer. Anaerocult®-systemet er baseret ...
MSDS Tilbud
Leverandør: THERMO FISHER DIAGNOSTICS GMBH
Beskrivelse: The jar is designed for use with AnaeroGen/CampyGen sachet and will hold up to 12 plates.
Varenummer: (710-1964)
Leverandør: BIOTOOL AG
Beskrivelse: The PetriSphere anaerobic gas system is suitable for the culturing of anaerobic, microaerophilic and capnophilic bacteria in common anaerobic jars. At...
UOM: 1 * 1 Items
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: Sample manipulation within a secure anaerobic environment is now as easy as working aerobically at the laboratory bench.
Varenummer: (710-0912)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The Whitley A85 Anaerobic Workstation has been created to enable both research and clinical scientists to carry out microbial manipulation within a se...
UOM: 1 * 1 Items
Varenummer: (DOWIA08400)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The A135 is ideal for studies where a variety of equipment needs to be used within an anaerobic environment.
UOM: 1 * 1 Items
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: These anaerobic jars are robustly constructed in stainless steel and are simple to use.
Varenummer: (DOWIA07400)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The A95 Workstation has a huge capacity of up to 1120×90mm Petri dishes, depending upon the way you choose to configure your working and incubation ar...
UOM: 1 * 1 Items
Leverandør: schuett-biotec GmbH
Beskrivelse: For the cultivation of anaerobic or microaerophilic microorganisms in a defined, rapidly developing and optimal gas atmosphere. Racks (optional): For ...
Leverandør: BEHR
Beskrivelse: behrotest® serial extraction devices are the economical and user-friendly solution for classic Soxhlet and fat extraction. With behr hydrolysis units ...
Varenummer: (BEHRB00217739)
Leverandør: BEHR
Beskrivelse: behrotest® serial extraction devices are the economical and user-friendly solution for classic Soxhlet and fat extraction.
With behr hydrolysis u...
UOM: 1 * 1 Items
Varenummer: (DOWIA09020)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The A155 is the largest workstation in the Whitley Workstation range – 350mm taller than the A55.
UOM: 1 * 1 Items
Varenummer: (DOWIA09000)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The A25 is a truly compact workstation equipped with many unique features to ensure easy and efficient use and to guarantee that the very best anaerob...
UOM: 1 * 1 Items
Varenummer: (DOWIA07760)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The Whitley A55 has a chamber size twice the width of an A35, with a huge working and incubation area and airlocks at both ends.
UOM: 1 * 1 Items
Varenummer: (710-0913)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The Whitley A45 Anaerobic Workstation is 50% larger than a Whitley A35 and has three ports to provide convenient access to the entire incubation and w...
UOM: 1 * 1 Items
Varenummer: (710-1698)
Leverandør: DON WHITLEY SCIENTIFIC LTD
Beskrivelse: The Whitley jar gassing system provides perfect conditions for growing anaerobes in jars in just 2 minutes, and microaerophiles in 15 seconds.
UOM: 1 * 1 Items
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 16
no targeter for Bottom