3-Amino-2,2-dimethyl-1-propanol 97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
ACRO457430250EA
2050
DKK
ACRO457430250
3-Amino-2,2-dimethyl-1-propanol 97%
3-Amino-2,2-dimethyl-1-propanol
Formel:
C₅H₁₃NO Smeltepunkt: 70 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00059847 CAS-nummer: 26734-09-8 |
Specifikationer Testresultater
Appearance | Colorless to white to light yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...