Du Søgte efter: Elektrodeholdere
15  results were found
SearchResultCount:"15"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Varenummer: (241-0114)
Leverandør: VWR Collection
Beskrivelse: Grey iron stand for two electrodes.
UOM: 1 * 1 Items
Varenummer: (662-1169)
Leverandør: VWR Collection
Beskrivelse: Electrode arm moves smoothly in almost any direction, holding up to four electrodes plus one temperature sensor safely and securely in selected positi...
UOM: 1 * 1 Items
Varenummer: (BELAH185090000)
Leverandør: Bel-Art Products, a Part of SP
Beskrivelse: This handy rack made from PP holds eight test tubes for storing and conditioning pH and ion electrodes.
UOM: 1 * 1 Items
Varenummer: (662-1155)
Leverandør: SI Analytics
Beskrivelse: Plastic receptacle with squeezing ring sealing and bayonet fastener for electrodes with 12 mm diameter.
UOM: 1 * 1 Items
Varenummer: (662-0412)
Leverandør: WTW
Beskrivelse: Bordstativ med fleksibel elektrodeholder. Velegnet til pH-komboelektroder og temperatursensorer.
UOM: 1 * 1 Items
Varenummer: (662-1137)
Leverandør: Thermo Orion
Beskrivelse: This electrode stand with 360° motion and cable management system holds up to five electrodes with 12 mm diameter. With weighted base, swing arm and e...
UOM: 1 * 1 Items
Varenummer: (HACH9260300)
Leverandør: Hach
Beskrivelse: Electrode storage bottle is used to store pH electrodes.
UOM: 1 * 1 Items
Leverandør: Thermo Orion
Beskrivelse: These electrode holders can be used with the Star A series portable meter armor and storage sleeve to protect, transport and store sensors in the fiel...
Varenummer: (EUTEECPHELSTDC)
Leverandør: EUTECH
Beskrivelse: This electrode stand with swivel arm allows convenient hands-free operation. Holds up to four standard size electrodes and one ATC probe.
UOM: 1 * 1 Items
Varenummer: (ORIOSTARA-ELHD)
Leverandør: Thermo Orion
Beskrivelse: Use the Thermo Scientific™ Orion™ Star A series electrode holder with a meter-attached or swing arm electrode stand to organize your sensors for easie...
UOM: 1 * 1 Items
Varenummer: (ORIOSTARA-BEA)
Leverandør: Thermo Orion
Beskrivelse: This meter-attached electrode stand can be used to easily place sensors in samples.
UOM: 1 * 1 Items
Varenummer: (ORIO081045)
Leverandør: Thermo Orion
Beskrivelse: These protective guards provide additional protection and weight for conductivity cells and DO probes.
UOM: 1 * 1 Items
Varenummer: (ORIOSTARA-HB)
Leverandør: Thermo Orion
Beskrivelse: This weighted base can be used with the meter-attached electrode stand for independent stand use and compatibility with any meter system.
UOM: 1 * 1 Items
Varenummer: (ORIOSTARA-AR)
Leverandør: Thermo Orion
Beskrivelse: This portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electr...
UOM: 1 * 1 Items
Varenummer: (ORIOSTARA-CS)
Leverandør: Thermo Orion
Beskrivelse: This hard-sided field case can be used to protect, transport and store measuring equipment in the field.
UOM: 1 * 1 Items
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 15 of 15
no targeter for Bottom