VWR®, Slider Zip Bags
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
129-1200EA 1170 DKK
129-1200 129-1201 129-1198 129-1199 129-1202 129-1203
VWR®, Slider Zip Bags
Poser Opbevaringsposer
VWR® slider zip bags, in LDPE, transparent, with blue slider. For the packing and storage of goods.
  • Can be used from −10 to +65 °C
  • Do not contain any ingredients of animal origin
Certificeringer: Manufactured under ISO 9001:2015 standard.
Emballering: 50 pcs/outer bag, 10 outer bags/case.