Whole blood, psoriatic arthritis (PA)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HCMSPB000F-2-PAEA 0 DKK
HCMSPB000F-2-PA
Whole blood, psoriatic arthritis (PA)
Whole blood is used for researching the causes, diagnosis, supportive care and treatment of Psoriatic arthritis (PA). Collected from IRB-consented donors diagnosed with psoriatic arthritis.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Advarsel: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.