High performance undercounter refrigerators, TSX
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HERATSX505SVEA 37510 DKK
HERATSX505SV
High performance undercounter refrigerators, TSX
Designed to preserve the environment, the Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other research and medical products. Powered by V-drive, the TSX Series delivers low energy, decreased noise levels and improved performance and user experience. These undercounter refrigerators are quiet and feature an improved user interface, automated locking and increased storage capacity and reliability.
  • Redundant cooling technology protects the storage while delivering 37% less energy (1,7 kWh/day), 20 times less noise 35 dBA) and twice the storage capacity
  • No compressor
  • Datalogging with USB download funtionality
Compressor-free V-drive featuring STeM (synchronized temperature management):
Dual refrigeration system combines the benefits of cold wall and fan-assisted recovery
Actively monitors the cabinet to maintain the desired temperature conditions
Constant monitoring and adjustment during periods of door openings keeps temperatures stable without temperature fluctuation found in models with compressors that cycle on/off
This tight internal air circulation eliminates the risk of freezing which can ruin materials like vaccines
Certificeringer: UL, CE, REACH, RoHS