Anti-PECAM1 Mouse Monoclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-872EA 3590 DKK
PRSI49-872
Anti-PECAM1 Mouse Monoclonal Antibody
Antistoffer
PECAM1 antibody can be used in Western Blot starting at 1:2000, and immunohistochemistry starting at 2.5 μg/mL.

Affinity Chromatography

PBS, pH 7.2, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PECAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR