Anti-TPSAB1 Mouse Monoclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-867EA 4620 DKK
PRSI49-867
Anti-TPSAB1 Mouse Monoclonal Antibody
Antistoffer
Tryptase antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL. This product requires protein digestion pre-treatment of paraffin sections e.g. pronase. Results may also be obtained with this produst using HIER. Sodium citrate buffer pH 6.0 is recommended for this purpose.

Protein G Column

PBS, pH 7.4, 0.09% sodium azide.

Store Tryptase antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles. Store undiluted.

Type:
Antigen: TPSAB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Dog, Human, Monkey, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR